missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDGF Antibody (OTI8C7), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70882DL594
This item is not returnable.
View return policy
Description
HDGF Monoclonal antibody specifically detects HDGF in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| HDGF | |
| Monoclonal | |
| DyLight 594 | |
| FLJ96580, hepatoma-derived growth factor, hepatoma-derived growth factor (high-mobility group protein 1-like), High mobility group protein 1-like 2, HMG-1L2, HMG1L2DKFZp686J1764 | |
| Full length recombinant protein of human HDGF (NP_004485) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Cell Biology | |
| 3068 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI8C7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction