missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDHD1 Monoclonal Antibody (OTI6G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502644S
This item is not returnable.
View return policy
Description
HDHD1 Monoclonal antibody specifically detects HDHD1 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HDHD1 | |
| Monoclonal | |
| 0.57 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14457, Q91XJ1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 114558, 480513, 8678 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI6G3 | |
| Unconjugated | |
| BECN1 | |
| 1700121L12Rik; 5'-PsiMPase; DXF68S1E; FAM16AX; family with sequence similarity 16, member A, X-linked; GS1; haloacid dehalogenase-like hydrolase domain containing 1; haloacid dehalogenase-like hydrolase domain containing 1A; haloacid dehalogenase-like hydrolase domain-containing protein 1; Haloacid dehalogenase-like hydrolase domain-containing protein 1A; haloacid dehalogenase-like hydrolase domain-containing protein 1A-like; Hdhd1; Hdhd1a; Protein GS1; pseudouridine 5'-phosphatase; pseudouridine-5'-monophosphatase; Pseudouridine-5'-phosphatase; Pudp | |
| Full length human recombinant protein of human BECN1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction