missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDHD1A Antibody (OTI7A2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71368PCP
This item is not returnable.
View return policy
Description
HDHD1A Monoclonal specifically detects HDHD1A in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| HDHD1A | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| PUDP | |
| Full length recombinant protein of human HDHD1A (NP_036212) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI7A2 | |
| Flow Cytometry | |
| DXF68S1E5′-PsiMPase, EC 3.1.3.n6, family with sequence similarity 16, member A, X-linked, GS1FAM16AX, haloacid dehalogenase-like hydrolase domain containing 1, haloacid dehalogenase-like hydrolase domain containing 1A, Haloacid dehalogenase-like hydrolase domain-containing protein 1, Haloacid dehalogenase-like hydrolase domain-containing protein 1A, HDHD1Apseudouridine-5′-monophosphatase, Protein GS1 | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 8226.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction