Learn More
MST1/STK4 Antibody (06), HRP , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06387H
Product Code. 18627136
  • € 423.00 / 0.10mL
Description
MST1/STK4 Monoclonal antibody specifically detects MST1/STK4 in Human samples. It is validated for ELISA, ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MST1/STK4
Monoclonal
HRP
Mouse
Protein A purified
RUO
Human
Purified
ELISA
06
PBS
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MST1/STK4 (Q13043-1; Glu2-Phe487).
0.1 mL
Primary
Store at 4°C in the dark.