missing translation for 'onlineSavingsMsg'
Learn More

HDHD2 Antibody (OTI4G3), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-72259PCP

Product Code. 18145943

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HDHD2 Monoclonal specifically detects HDHD2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDHD2
Monoclonal
PerCP
PBS with 0.05% Sodium Azide
HDHD2
Full length recombinant protein of human HDHD2 (NP_115500) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Flow Cytometry
OTI4G3
Flow Cytometry
DKFZp564D1378, DKFZP564D1378,3110052N05Rik, haloacid dehalogenase-like hydrolase domain containing 2, haloacid dehalogenase-like hydrolase domain-containing protein 2
Mouse
Affinity Purified
Lipid and Metabolism
84064.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.