missing translation for 'onlineSavingsMsg'
Learn More

Pigment Red 22, Thermo Scientific™

Product Code. 17505545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17505545 5 g 5g
17595535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17505545 Supplier Thermo Scientific Acros Supplier No. 468500050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6448-95-9
Molecular Formula C24H18N4O4
Molecular Weight (g/mol) 426.43
MDL Number MFCD00071989
InChI Key HVBDWKMAUMVFIY-QYQHSDTDSA-N
IUPAC Name (4Z)-4-[2-(2-methyl-5-nitrophenyl)hydrazin-1-ylidene]-3-oxo-N-phenyl-3,4-dihydronaphthalene-2-carboxamide
SMILES CC1=CC=C(C=C1N\N=C1/C(=O)C(=CC2=CC=CC=C12)C(=O)NC1=CC=CC=C1)[N+]([O-])=O

Specifications

Infrared Spectrum Conforms
Loss on Drying ≤2 %
Packaging Glass Bottle
Certificate of Analysis Available
Color Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Pigment Red 22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.