missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDLBP Recombinant Rabbit Monoclonal Antibody (23GB3970), Invitrogen™
GREENER_CHOICE

Product Code. p-200088497
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089292 100 μL 100µL
30088905 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089292 Supplier Invitrogen™ Supplier No. MA552648

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

High density lipoprotein-binding protein, also known as vigilin, is a 110-kD protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDLBP
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3970
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HDLBP
Gene Accession No. Q00341, Q8VDJ3, Q9Z1A6
Gene Alias 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDL binding protein; HDL-binding protein; HDLBP; Hemopexin; high density lipoprotein (HDL) binding protein; high density lipoprotein binding protein; high density lipoprotein binding protein (vigilin); high density lipoprotein-binding protein; Hpx; Hpxn; hx; lipoprotein-binding protein; PRO2900; VGL; vigilin
Gene Symbols HDLBP
Host Species Rabbit
Immunogen A synthesized peptide derived from human HDLBP (70-100AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110611, 3069, 64474
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.