Learn More
Verenigde Kunststof Bedrijven™ HDPE Barrel
Product Code. 10597554
Change view
Click to view available options
Capacity (Metric):
30 L
60 L
220 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10597554 220 L Each
10257354 30 L Each
10696625 60 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10597554 Supplier Verenigde Kunststof Bedrijven™ Supplier No. 608815
In Stock
Add to basket
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Verenigde Kunststof Bedrijven™ HDPE Barrel is designed for storage, handling and transportation of products manufactured by the chemical, petrochemical and food industries.
  • HDPE UN approved Lid barrels
  • Removable black Lid
  • Metal fastener
  • For transport and storage Chemical, Petrochemical, Food Industries
  • Complete Draining
  • enabling re-use or recycling
  • Does not age or corrode
  • Easy to Stack and pallatise
Specifications
Certifications/Compliance UN Approved
Product Type Barrel
Closure Type Removable Lid
Capacity (Metric) 220 L
Color Blue
Material HDPE
Shape Round
Length (Metric) 587 mm
Height (Metric) 967 mm