Learn More
Karl Hecht™ Funnel, Polyethylene
Product Code. p-8004059
Change view
Click to view available options
Diameter (Metric)Top:
6mm
8mm
16mm
14cm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top unitSize
10033741 6mm Each
10238061 8mm Each
10115591 14cm Each
10400202 16mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10033741 Supplier Karl Hecht™ Supplier No. 49050003
Multi-use plastic funnel. Karl Hecht™ Funnel, Polyethylene facilitates pouring and reduces spilling risks.
  • Polyethylene construction.
  • Facilitates pouring operations.
  • Reduces the risk of spilling.
Specifications
Diameter (Metric)Top 6mm
Material Polyethylene
For Use With (Application) Filtering Of Samples
Length (Metric) Stem 40mm
Quantity 10/Pk.
Product Type Funnel