Learn More
Huenersdorf™ HDPE Industrial Bucket
Click to view available options
Capacity (Metric):
10.5 L
14 L
17 L
Unit Size:
Each
Color:
Natural
Description:
Round
Description
- Square
- Natural
- Acid-resistant polyethylene (HDPE)
- Embossed scale and plastic handle Industrial bucket for safe handling
Specifications
Specifications
| Product Type | Industrial Bucket |
| Capacity (Metric) | 14 L |
| Material | HDPE |
| Dimensions | 292 x 292 x 292 mm |
| Height (Metric) | 292 mm |
| Length (Metric) | 292 mm |
| Shape | Square |