Learn More
Kartell™ HDPE Industrial Scoop
Click to view available options
Capacity (Metric):
1500 mL
Unit Size:
Pack of 25
Description
- Constructed of HDPE
- Designed for heavy industrial use
- Features a round shaped design to facilitate complete transfer of sampling material
- Suitable for foodstuff
Specifications
Specifications
| Quantity | 1 Ea. |
| Capacity (Metric) | 1500 mL |
| Color | Neutral |
| Product Type | Scoop |
| For Use With (Application) | Suitable for foodstuff |
| Material | HDPE |
| Length (Metric) | 400 mm |