Learn More
Kautex™ High Density Polyethylene Medium Mouth Bottles
Manufactured with HDPE for stability, strength and opacity. Kautex™ High Density Polyethylene Medium Mouth Bottles have an embossed graduated scale for permanent accurate measurement.
Brand:  Kautex™ 2000770070
Additional Details : Weight : 0.43200kg
 View more versions of this product
Product Code. 10596293
  • € 26.50 / Pack of 10
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Multi-purpose bottles for laboratories and industry
  • Embossed contents scale
  • Easy to fill and clean, leak-proof
  • PP screw closure
Application:
  • dispensing
  • sending samples
  • mailing
  • transport
  • storage
Specifications
Specifications
Polypropylene
161 mm
Screw Cap
500 mL
30.8 mm
HDPE
1 Ea.
1 Ea.
Yes
Bottle
77 mm
40 mm
Natural
Packaging, Sending Samples, Transport, Mailing and Storage
10/Cs.
Bottle
Yes
Round