Learn More
Kautex™ HDPE 308/310 Series Narrow Neck Bottle
Product Code. 10346211
Click to view available options
Capacity (Metric):
0.5 L
1 L
2.5 L
Color:
Black
Natural
Unit Size:
Each
Pack of 120
Pack of 26
Pack of 64
Product Code. 10346211
Brand: Kautex™ 2000084464
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use this Kautex™HDPE 308/310 Series Narrow Neck Bottle for packaging, sending samples, mailing and storage. Suitable for filling and transport of hazardous liquids, it features a recessed labeling field with sizes 1000 and 2500 mL. and is UN approved for liquids.
  • Optimized handle for safe and comfortable handling.
  • Recessed labelling field with sizes 1000 and 2500 mL.
  • GL 45 neck, three different types of closures available
  • leakproof
  • UN approved for liquids
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 0.5 L
Color Natural
Diameter (Metric) Inner Neck 28.7 mm
For Use With (Application) Packaging, Sending Samples, Mailing, Storage
Material HDPE
Neck Finish Narrow
Certifications/Compliance UN 1H1/X1.9/250
Mouth Narrow
Height (Metric) 176.5 mm
Product Type Narrow Neck Bottle
Diameter (Metric) 77 mm