Learn More
Kautex™ HDPE Round Bottle
Product Code. 10285281
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Inner Neck:
17 mm
19.4 mm
Unit Size:
Each
Pack of 148
Pack of 72
Pack of 792
Pack of 900
Product Code. 10285281
Brand: Kautex™ 2000073086
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Kautex™ HDPE Round Bottle for many bottling needs, including cosmetic products, e.g. emulsions, shampoos, shower gels, creams and as a refill packaging. Order PP hinged closure separately;
  • Natural color
  • Stable and handy shape
  • PP hinged closure ordered separately
  • Hinged lid closure for single-handed use
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Diameter (Metric) Inner Neck 19.4 mm
For Use With (Application) Packaging, Storage and Transport
Material HDPE
No. per Case 148/Cs.
No. per Pack 1 Ea.
Packaging Bottle
Quantity 1 Ea.
Screw Color White
Screw Material Polypropylene
Closure Color White
Closure Included No
Shape Round
Product Type Bottle
Closure Material Polypropylene
Closure Size 22 mm
Diameter (Metric) 70 mm
Closure Type Hinged Lid