Learn More
Kautex™ HDPE Round Bottle
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Inner Neck:
17 mm
19.4 mm
Unit Size:
Each
Pack of 148
Pack of 72
Pack of 792
Pack of 900
Description
- Natural color
- Stable and handy shape
- PP hinged closure ordered separately
- Hinged lid closure for single-handed use
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Diameter (Metric) Inner Neck | 17 mm |
| For Use With (Application) | Packaging, Storage and Transport |
| Material | HDPE |
| No. per Case | 729/Cs. |
| No. per Pack | 1 Ea. |
| Packaging | Bottle |
| Quantity | 1 Ea. |
| Screw Color | White |
| Screw Material | Polypropylene |
| Show More |