Learn More
Sampling Systems™ HDPE Thumb Tube Pipette
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
60 mL
Unit Size:
Pack of 50
Description
Key Features:
- Manufactured with HDPE
- Individually bagged
- FDA conforming materials
- BSE/TSE free
- Full batch traceability
- Available sterilized by gamma
Operation:
- Press thumb onto the tube until you are at the sampling depth
- Raise the thumb until levels equalise
- Press thumb onto the tube to seal the sample in
Specifications
Specifications
| Product Type | Thumb Tube Pipette |
| Material | HDPE |
| Capacity (Metric) | 60 mL |
| Disposable | Single-use |
| Sterility | Sterile |
| Certifications/Compliance | FDA approved CFR 177.1520, Conforms to EU regs 10/2011 |
| Length (Metric) | 1000 mm |