missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HEATR5B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15797671 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15797671 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15797671 Supplier Invitrogen™ Supplier No. PA559827

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VPPPVSAALQ GIKSIVTLSM AKTEAGVQKQ WTALIRSTLA CILEYSQPED SVPTPDEVSM LTAIALFLWS ASNEIIGVQS LQNGCMNRF Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

HEATR5B gene ontology annotations related to this gene include endocytosis; protein localization; retrograde transport, endosome to Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEATR5B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEATR5B
Gene Accession No. Q9P2D3
Gene Alias 2010013B10Rik; A230048G03Rik; AV071430; D330050P16Rik; HEAT repeat containing 5B; HEAT repeat-containing protein 5B; Heatr5b; Kiaa1414; mKIAA1414
Gene Symbols HEATR5B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEATR5B. Recombinant protein control fragment (Product #RP-95950).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54497
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.