missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEATR9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570616
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 85%; Guinea Pig: 79%; Human: 100%; Rat: 100%.
HEATR9 gene ontology annotations related to this gene include hematopoietic progenitor cell differentiation.
Specifications
| HEATR9 | |
| Polyclonal | |
| Unconjugated | |
| HEATR9 | |
| C11H17orf66; C17orf66; C9H17orf66; Gm11435; HEAT repeat containing 9; HEAT repeat-containing protein 9; Heatr9; OTTMUSG00000000990; Protein HEATR9; RGD1559875 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 256957 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A2RTY3 | |
| HEATR9 | |
| synthetic peptide directed towards the middle region of human HEATR9 (aa 473-522). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction