missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109303
This item is not returnable.
View return policy
Description
Positive Control: SiHa, HL-60, MG-63.
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
Specifications
| HEC1 | |
| Polyclonal | |
| Unconjugated | |
| NDC80 | |
| 2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10403 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O14777 | |
| NDC80 | |
| Recombinant protein within Human HEC1 aa 300-505. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction