missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HECW1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-93645-25ul
This item is not returnable.
View return policy
Description
HECW1 Polyclonal specifically detects HECW1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HECW1 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q76N89 | |
| HECW1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:STEPESAQIQDSPMNNLMESGSGEPRSEAPESSESWKPEQLGEGSVPDGPGNQSIELSRPAEEAAVITEAGDQGMVSVGPEGAGELLAQVQKDIQPAPSAEELAEQLDLGEEASALLLEDGEAPAS | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C2 and WW domain-containing protein 1, HECT type E3 ubiquitin ligase, HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1, NEDD4-like E3 ubiquitin-protein ligase 1, NEDL1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23072 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction