missing translation for 'onlineSavingsMsg'
Learn More

HEG1 Antibody (HEG1/9445), Novus Biologicals™

Product Code. p-200112517 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30290010 100 μg 100µg
30289756 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30290010 Supplier Novus Biologicals Supplier No. NBP344618100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HEG1 Monoclonal antibody specifically detects HEG1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEG1
Applications Immunohistochemistry
Classification Monoclonal
Clone HEG1/9445
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation 10 mM PBS with 0.05% BSA
Gene Alias HEG homolog 1 (zebrafish), MST112, MSTP112
Host Species Mouse
Immunogen Recombinant fragment (around aa1-400) of human HEG1 protein (Uniprot: Q9ULI3). Exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57493
Target Species Human
Content And Storage Store at 4⪚C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.