Learn More
Heidolph™ Hei-VAP Twin VAP Package, EU-plug
Product Code. 17383330 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
EU-plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
17383330 EU-plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17383330 Supplier Heidolph™ Supplier No. 5742930000
Only null left
Add to basket
Add to basket
The Hei-VAP Twin VAP Package features a motor lift, vacuum pump control, chiller, and tubing set.
  • 2 x Hei-VAP Expert Control with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Valve Control
  • 2 x Vacuum valve
  • Chiller Hei-CHILL 1200
  • 2 x Tubing set
  • Thermal fluid Kyro 30 (20 L)
  • 2 x Non-return valve
  • 2 x Y-Piece
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type EU-plug