Description
- 2 x Hei-VAP Expert Control with motor lift and vertical glassware set G3
- Vacuum pump Rotavac Valve Control
- 2 x Vacuum valve
- Chiller Hei-CHILL 1200
- 2 x Tubing set
- Thermal fluid Kyro 30 (20 L)
- 2 x Non-return valve
- 2 x Y-Piece
Specifications
Specifications
| Type | Motor Lift Rotary Evaporator |
| Plug Type | EU-plug |