Learn More
Horst Heizpatrone, DxL= 12,5 x 80 mm, 250 W
Brand:  Horst 514081
Heizpatrone, DxL= 12,5 x 80 mm, 250 W
Product Code. 10780023
  • ONLINE EXCLUSIVE PRICE
    € 69.25
    € 61.94 / Each
    Save € 7.31
    10% Off
Only null left
Estimated Shipment: 28-01-2026