Learn More
Thermo Scientific™ Hektoen Enteric Agar/DCA Leifson Agar Biplate
Description
Isolate and differentiate Shigella and Salmonella spp. from enteric pathological specimens with this convenient bi-plate.
Isolate and differentiate Shigella and Salmonella spp. from enteric pathological specimens while simultaneously producing larger and numerous colonies of Shigella spp. that can easily be picked off and emulsified in saline for slide agglutination tests with Thermo Scientific™ Oxoid™ Hektoen Enteric Agar/DCA Leifson Agar Biplate. The convenient biplate format allows simultaneous isolation and detection of Shigella and Salmonella spp. from stool specimens.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
Hektoen Enteric Agar/DCA Leifson Agar Biplate allows simultaneous isolation and detection of Shigella and Salmonella spp.
- Easy-to-read: on Hektoen Enteric Agar shigellae form green moist raised colonies, while salmonellae form blue-green colonies, and other coliforms form salmon-pink to orange colonies surrounded by a zone of bile precipitation.
- Selective: selective nature of Hektoen Enteric Agar and DCA Leifson Agar improves the recovery of salmonellae and shigellae.
- Ready-to-use: convenience of prepared media
- Space saving: bi-plate format
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Color | Olive to ochre yellow, transparent/ Salmon orange, transparent |
| Content And Storage | 2° to 12°C, in the dark |
| Description | Hektoen Enteric Agar/ DCA Leifson Agar Biplate |
| Format | Biplate |
| Manufacturing Origin | Germany |
| Product Type | Agar Plate |
| Fill Weight | 17g + 5% |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10 Pack |