Learn More
Invitrogen™ Helicobacter pylori 54 kDa Monoclonal Antibody (6644)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547577
Description
Specificity: Reactive with the 54 kDa (flagellin) of H. pylori. Non-reactive with (ICC/IF): Giardia, Cryptosporidium, Enterococcus, E. coli sp., Salmonella sp., Shigella sp., Campylobacter & Vibrio. Best Pairs for Antigen Detection (capture/conjugate): Mycoplasma pneumoniae Polyclonal antibody (Product # MA5-18231)/Helicobacter pylori 54 kDa Monoclonal antibody (Product # MA5-47577); Helicobacter pylori 54 kDa Monoclonal antibody (Product # MA5-47578)/Helicobacter pylori 54 kDa Monoclonal antibody (Product # MA5-47577). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
Specifications
| Helicobacter pylori 54 kDa | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2a |
| ELISA, Immunocytochemistry | |
| 6644 | |
| Unconjugated | |
| H. pylori | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.