missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Helicobacter Pylori HP-NAP Recombinant Rabbit Monoclonal Antibody (RM413)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544570
This item is not returnable.
View return policy
Description
This antibody reacts to Histone H2A.X acetylated at Lysine 5 (K5ac). No cross reactivity with non-modified Lysine 5 or other acetylated Lysines in histone H2A.
In H. pylori, the Helicobacter pylori neutrophil-activating protein (HP-NAP) is a virulence factor that attracts and activates neutrophils, and promotes their endothelial adhesion and the production of oxygen radicals and chemokines, including CXCL8, CCL3 and CCL4.
Specifications
| Helicobacter Pylori HP-NAP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Proteins purified from Helicobacter pylori which correspond to the full length protein. | |
| 100 μL | |
| Primary | |
| -20°C | |
| Liquid |
| ELISA | |
| RM413 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction