missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helicobacter Pylori Mouse anti-Bacteria, Clone: HP/1336, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07674-100ug
This item is not returnable.
View return policy
Description
Helicobacter Pylori Monoclonal specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Helicobacter Pylori | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1 to 2 μg/million cells, ELISA 1-5 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| H. pylori | |
| Total sonicate of Helicobacter pylori | |
| 100 μg | |
| Primary | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| HP/1336 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| H. pylori | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction