Learn More
Invitrogen™ Helicobacter Pylori OMP Recombinant Rabbit Monoclonal Antibody (RM410)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544572
This item is not returnable.
View return policy
Description
This antibody may also react to bovine or rat alpha smooth muscle Actin, as predicted by immunogen homology.
Outer membrane proteins (OMPs) ofH. pyloriplay a crucial role in the host pathogen interaction, virulence and pathogenesis. When OMPs interact with the host immune cells, OMPs stimulate the production of pro-inflammatory molecules that may result in an uncontrolled inflammation leading to the transformation of a normal cell to a cancerous cell. The Helicobacter outer membrane family of OMPs inH. pyloriconsists of four members (HomA, B, C and D).
Specifications
| Helicobacter Pylori OMP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Proteins purified from Helicobacter pylori which correspond to the full length protein. | |
| 100 μL | |
| Primary | |
| -20°C | |
| Liquid |
| ELISA | |
| RM410 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |