missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helios Antibody (22F6), Allophycocyanin, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP2-37723APC
This item is not returnable.
View return policy
Description
Helios Monoclonal specifically detects Helios in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| Helios | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| IKZF2 | |
| Peptide coresponding to the amino acids 51-107 of Helios. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 22807 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 22F6 | |
| Flow Cytometry | |
| HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
| Armenian Hamster | |
| Protein A purified | |
| RUO | |
| Primary | |
| The armenian hamster monoclonal antibody 22F6 recognizes Helios, a transcription factor expressed in some hematopoietic stem cells, and at high levels in thymic-derived regulatory T cells. | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction