missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helios Antibody (736440), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB73092MFV500
This item is not returnable.
View return policy
Description
Helios Monoclonal antibody specifically detects Helios in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Helios | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 736440 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
| E. coli-derived recombinant human Helios, Met1-Gln97, Accession # Q9UKS7 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 22807 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction