missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HELIOS Monoclonal Antibody (22F6), Brilliant Ultra Violet™ 563, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
€ 213.00 - € 429.00
Specifications
| Antigen | HELIOS |
|---|---|
| Clone | 22F6 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17806256
![]() |
Invitrogen™
365-9883-42 |
100 Tests |
€ 429.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17846246
|
Invitrogen™
365-9883-41 |
25 Tests |
€ 213.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.Specifications
| HELIOS | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| A730095J18Rik; ANF1A2; Helios; IKAROS family zinc finger 2; IKAROS family zinc finger 2 (Helios); ikaros family zinc finger protein 2; Ikzf2; Zfpn1a2; zinc finger DNA binding protein Helios; zinc finger protein Helios; zinc finger protein, subfamily 1A, 2 (Helios); ZNF1A2; Znfn1a2 | |
| IKZF2 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 22F6 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 563 | |
| Armenian Hamster | |
| Canine, Human, Mouse | |
| P81183, Q9UKS7 | |
| 22779, 22807, 488510 | |
| IgG | |
| Affinity chromatography | |
| IKZF2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title