Learn More
Invitrogen™ HELT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557564
Description
Immunogen sequence: SDKLKERKVS RLSPNGTCAL VVEASDSPTR HLGGPMAGKC PHGTLSVEES RVRT Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 30%.
Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGCG-3'.
Specifications
| HELT | |
| Polyclonal | |
| Unconjugated | |
| HELT | |
| A830086M02; bHLH transcription factor megane; bHLHb44; Hairy and enhancer of split-related protein HELT; HCM1228; Helt; helt bHLH transcription factor; Helt-ps1; HES/HEY-like transcription factor; HES/HEY-like transcription factor, pseudogene 1; Hesl; Heslike; Hey-like transcription factor; Hey-like transcriptional repressor; megane; Megane bHLH factor; mesencephalic GABAergic neurons enhancer of split related protein; Mgn; Protein Hes-like; Protein megane; RGD1564073 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 391723 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NFD8 | |
| HELT | |
| Recombinant protein corresponding to Human HELT. Recombinant protein control fragment (Product #RP-96752). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.