missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HELT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24730-1-AP-150UL
This item is not returnable.
View return policy
Description
Transcriptional repressor which binds preferentially to the canonical E box sequence 5′-CACGCG-3′.Specifications
| HELT | |
| Polyclonal | |
| Unconjugated | |
| HELT | |
| HCM1228, HELT, HESL, Mgn | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234219, 391723, 498633 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A6NFD8, Q7TS99 | |
| HELT | |
| HELT Fusion Protein Ag20618 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction