missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies HemaTox™ Erythroid Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- Screen large numbers of novel drug compounds for their effect on the proliferation and erythroid-specific differentiation of human hematopoietic stem and progenitor cells (HSPCs)
- Compared to using the colony-forming unit (CFU) assay, the liquid culture-based HemaTox assay is more flexible: you can add test compounds at various points throughout the culture and measure their effects on more primitive or more mature progenitor cells
- The HemaTox Erythroid Kit promotes the proliferation of CD34+ HSPCs and their differentiation into cells expressing erythroid markers such as CD71 and CD235a
- Users can choose the readout method to quantify the response and estimate the IC50 and IC90 of each compound tested
- Test up to 160 conditions in triplicate per kit in 5 x 96-well plates
- This kit may be used on its own or in combination with HemaTox Myeloid Kit or HemaTox Megakaryocyte Kit to assess lineage-specific drug toxicity in parallel
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells, Erythroid Cells |
| For Use With (Application) | Cell Culture; Toxicity Assay |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction