missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Heme oxygenase 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549705
This item is not returnable.
View return policy
Description
The antibody detects endogenous level of total Heme oxygenase 2 protein.
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family. Several alternatively spliced transcript variants encoding three different isoforms have been found for this gene.
Specifications
| Heme oxygenase 2 | |
| Polyclonal | |
| Unconjugated | |
| HMOX2 | |
| heme oxygenase (decycling) 2; heme oxygenase 2; heme oxygenase-2 non-reducing isoform; HMOX 2; HMOX2; HO 2; HO2; HO-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3163 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P30519 | |
| HMOX2 | |
| Fusion protein corresponding to a region derived from 82-316 amino acids of human heme oxygenase (decycling) 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction