missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Heme oxygenase 2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA528334

Product Code. 13432447

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (88%), Rat (89%), Pig (87%), Rabbit (91%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family. Several alternatively spliced transcript variants encoding three different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Heme oxygenase 2
Polyclonal
Unconjugated
HMOX2
heme oxygenase (decycling) 2; heme oxygenase 2; heme oxygenase-2 non-reducing isoform; HMOX 2; HMOX2; HO 2; HO2; HO-2
Rabbit
Antigen affinity chromatography
RUO
3163
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.8 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P30519
HMOX2
Recombinant protein encompassing a sequence within the center region of human Heme Oxygenase 2. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.