missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEMGN Mouse anti-Human, Clone: UMAB169, liquid, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM870062
This item is not returnable.
View return policy
Description
HEMGN regulates the proliferation and differentiation of hematopoietic cells. Overexpression of HEMGN block the TPA-induced megakaryocytic differentiation in the K562 cell model. HEMGN may also prevent cell apoptosis through the activation of the nuclear factor-kappa B (NF-kB).Specifications
| HEMGN | |
| Monoclonal | |
| 0.5-1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BXL5 | |
| HEMGN | |
| Human recombit protein fragment corresponding to amino acids 260-484 of human HEMGN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| UMAB169 | |
| Unconjugated | |
| HEMGN | |
| CT155, EDAG, EDAG-1, NDR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55363 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction