missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemogen Antibody (OTI2D7), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72005V
This item is not returnable.
View return policy
Description
Hemogen Monoclonal specifically detects Hemogen in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Hemogen | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HEMGN | |
| Recombinant protein fragment corresponding to amino acids 260-484 of human Hemogen (NP_060907) produced in E.coli. | |
| 0.1 mL | |
| 55363.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2D7 | |
| Western Blot, Immunohistochemistry | |
| EDAGEDAG-1, Erythroid differentiation-associated gene protein, hemogen, Hemopoietic gene protein, NDR, Negative differentiation regulator protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction