missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin alpha Antibody, Alexa Fluor™ 350 , Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB110-41083AF350
This item is not returnable.
View return policy
Description
Hemoglobin alpha Polyclonal antibody specifically detects Hemoglobin alpha in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Hemoglobin alpha | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 350 | |
| alpha one globin, alpha-1 globin, alpha-1-globin, alpha-globin, CD31, hemoglobin alpha 1 globin chain, Hemoglobin alpha chain, hemoglobin alpha-1 chain, hemoglobin subunit alpha, hemoglobin, alpha 1, MGC126895, MGC126897 | |
| Human Hemoglobin | |
| 1 mL | |
| Signal Transduction | |
| 3039 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction