missing translation for 'onlineSavingsMsg'
Learn More

Hemoglobin beta Antibody (1G8) [DyLight 405] - Chimeric, Novus Biologicals Biologicals™

Product Code. 30522607 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522607 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522607 Supplier Novus Biologicals Supplier No. NBP343581V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Hemoglobin beta Monoclonal antibody specifically detects Hemoglobin beta in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemoglobin beta
Applications ELISA
Classification Monoclonal
Clone 1G8
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias beta globin chain, beta-globin, CD113t-C, HBBB, HBD, Hemoglobin beta chain, hemoglobin subunit beta, hemoglobin, beta
Host Species Rabbit
Immunogen The original antibody was generated by immunizing BALB/c mice with synthetic N-terminal glycopeptide (VHLTPEC) and KLH conjugate. (Uniprot# P68871)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3043
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.