missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin Goat anti-Human, HRP, Polyclonal , Abnova™
Goat polyclonal antibody raised against human Hemoglobin.
Brand: Abnova PAB29658.1mg
This item is not returnable.
View return policy
Specifications
| Hemoglobin | |
| Polyclonal | |
| Goat polyclonal antibody raised against human Hemoglobin. | |
| In 50mM HEPES (Stabilizer, 0.002% Thimerosal) | |
| Native purified human Hemoglobin. | |
| 1 mg | |
| Primary | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| HRP | |
| ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.5-5.0 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Goat | |
| Antigen Affinity Purification | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction