missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemopexin Antibody (036), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89686V
This item is not returnable.
View return policy
Description
Hemopexin Monoclonal antibody specifically detects Hemopexin in Human samples. It is validated for ELISA
Specifications
| Hemopexin | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 036 | |
| ELISA | |
| beta-1B-glycoprotein, FLJ56652, hemopexin, HX | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Hemopexin (NP_000604.1; Met 1-His 462). | |
| 0.1 mL | |
| Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases | |
| 3263 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction