missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 748.00
Specifications
| Antigen | Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18425332
|
Novus Biologicals
NBP1-92010-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18438101
|
Novus Biologicals
NBP1-92010 |
0.1 mL |
€ 748.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Polyclonal antibody specifically detects Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp547H098, EC 2.8.2, EC 2.8.2.-, FLJ25392, heparan sulfate 6-O-sulfotransferase 1, heparan-sulfate 6-sulfotransferase, HS6ST-1, HS6STheparan-sulfate 6-O-sulfotransferase 1, MGC116899, MGC116901 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QRYQYKRQLERREQRLRSREERLLHRAKEALPREDADEPGRVPTEDYMSHIIEKW | |
| Primary |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 9394 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title