missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparanase/HPSE Antibody (SR2148), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21871-100ul
This item is not returnable.
View return policy
Description
Heparanase/HPSE Monoclonal antibody specifically detects Heparanase/HPSE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Heparanase/HPSE | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR2148 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| EC 3.2, endo-glucoronidase, HEP, heparanase, Heparanase-1, Hpa1, HPAheparanase exon 9 and 10 deletion, HPR1, HPSE1heparanase-1, HSE1heparanase exon10-deletion | |
| A synthesized peptide derived from human Heparanase/HPSE (Uniprot #: Q9Y251) | |
| 100 μg | |
| Pancreatic Stem Cell Markers, Stem Cell Markers | |
| 10855 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu