missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis A Surface Antigen Antibody (BGN/B5B3), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62197AF594
This item is not returnable.
View return policy
Description
Hepatitis A Surface Antigen Monoclonal specifically detects Hepatitis A Surface Antigen in Virus samples. It is validated for ELISA.
Specifications
| Hepatitis A Surface Antigen | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Lysate from foetal kidney cells of rhesus monkey infected with extract from patient with active viral Hepatitis A. | |
| 0.1 mL | |
| NB100-62197 is specific for Hepatitis A surface antigen. | |
| Store at 4°C in the dark. |
| ELISA | |
| BGN/B5B3 | |
| ELISA | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Virus | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction