missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hepatitis B Virus Surface Ayw Monoclonal Antibody (Hs41)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA123078

Product Code. 11511440

  • € 534.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-23078 detects Hepatitis B Virus Surface Antigen (Ayw) in Human samples.

The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. The HBV surface protein antigens (HBsAg) are comprised of three carboxyl co terminal HBs proteins termed large (LHBs), middle (MHBs) and small (SHBs, also called major) protein. LHBs and MHBs also share the highly hydrophobic, repetitive, membrane spanning S domain. In addition, MHBs has a 55 amino acid region called preS2. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hepatitis B Virus Surface Ayw
Monoclonal
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Mouse
Protein G
RUO
Virus
Antibody
IgG1
ELISA
Hs41
Unconjugated
HBsAg; HBV; hepatitis b virus surface antigen; HepB
Recombinant HBsAg of ayw subtype.
100 μg
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documenti

Documenti

Certificati
Promozioni

Promozioni

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.