missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis B Virus X antigen Antibody (X36C), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22445IR
This item is not returnable.
View return policy
Description
Hepatitis B Virus X antigen Monoclonal specifically detects Hepatitis B Virus X antigen in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Hepatitis B Virus X antigen | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HBVgp3 | |
| Baculovirus expressed recombinant HBx. | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| X36C | |
| Western Blot, ELISA | |
| HBVgp3 X protein HBx, Hepatitis B Virus X Protein, Peptide X, pX, X protein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 944566 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction