missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus Antibody (BGN/1246/5G7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65251F
This item is not returnable.
View return policy
Description
Hepatitis C Virus Monoclonal antibody specifically detects Hepatitis C Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Hepatitis C Virus | |
| Monoclonal | |
| FITC | |
| HCV NS5, Hepatitis C virus non structural protein 5, Hepatitis C virus nonstructural protein 5, Hepatitis C Virus NS5 | |
| Synthetic peptide corresponding to the NS-5 antigen region of the native molecule. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| BGN/1246/5G7 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction