Learn More
Invitrogen™ Hepatitis C Virus Core Antigen Monoclonal Antibody (F66G)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17356
Description
The MA1-7356 antibody reacts with Hepatitis C virus core in viral and infected tissue samples. This antibody detects genotypes 1a,1b, and 2a. MA1-7356 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7356 antibody was raised against HCV Core protein and it is specific towards residues 29-39.
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
Specifications
| Hepatitis C Virus Core Antigen | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| F66G | |
| Unconjugated | |
| HCV core; HCVgp1; HepC; HepC core; polyprotein | |
| Hepatitis C virus core protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.