missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus Core Antigen Monoclonal Antibody (I81J)

Hepatitis C Virus Core Antigen Monoclonal Antibody (I81J)
Brand: Invitrogen MA17360
This item is not returnable.
View return policy
Description
Specifications
| Hepatitis C Virus Core Antigen | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA | |
| I81J | |
| Unconjugated | |
| HCV core; HCVgp1; HepC; HepC core; polyprotein | |
| Hepatitis C core protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction